1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)piperazine 95%
Supplier: Apollo Scientific
Warning
PC2076G-250MG PC2076G-1G
APOSPC2076G-250MGEA 42.2 EUR
APOSPC2076G-250MG APOSPC2076G-1G
1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)piperazine 95%
1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)piperazine
Formula: C₁₀H₁₁ClF₃N₃
Melting Pt: 63…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00173934
CAS Number: 132834-59-4