Cobalt(II) fluoride, anhydrous 99%
Supplier: Apollo Scientific
Danger
PC2080-100G PC2080-25G PC2080-1G
APOSPC2080-100GEA 168 EUR
APOSPC2080-100G APOSPC2080-25G APOSPC2080-1G
Cobalt(II) fluoride, anhydrous 99%
Cobalt (II) fluoride
Formula: CoF₂
Boiling Pt: 1400 °C
Melting Pt: 1200 °C
Density: 4.46 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010941
CAS Number: 10026-17-2
EINECS: 233-061-9
UN: 2923
ADR: 8+6.1,III