Copper(II) fluoride, anhydrous 99%
Supplier: Apollo Scientific
Danger
Synonyms: Cupric fluoride
PC2100-100G PC2100-25G PC2100-5G
APOSPC2100-100GEA 377 EUR
APOSPC2100-100G APOSPC2100-25G APOSPC2100-5G
Copper(II) fluoride, anhydrous 99%
Copper (II) fluoride
Formula: CuF₂
MW: 101.54 g/mol
Boiling Pt: 950
Melting Pt: 950 °C (dec.)
Density: 4.23 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010977
CAS Number: 7789-19-7
EINECS: 232-147-3
UN: 3260
ADR: 8 III
Merck Index: 12,02704