2,6-Dichloro-4-(trifluoromethyl)nicotinonitrile 97%
Supplier: Apollo Scientific
Warning
PC2101D-1G PC2101D-25G PC2101D-5G
APOSPC2101D-1GEA 21.9 EUR
APOSPC2101D-1G APOSPC2101D-25G APOSPC2101D-5G
2,6-Dichloro-4-(trifluoromethyl)nicotinonitrile 97%
2,6-Dichloro-4-(trifluoromethyl)nicotinonitrile
Formula: C₇HCl₂F₃N₂
MW: 241 g/mol
Melting Pt: 35…40 °C
Storage Temperature: Ambient
MDL Number: MFCD00084977
CAS Number: 13600-42-5
UN: 3439
ADR: 6.1 III