2-Nitro-4-(trifluoromethyl)benzonitrile 99%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Cyano-3-nitrobenzotrifluoride
PC2103-100G PC2103-500G PC2103-50G
APOSPC2103-100GEA 31.9 EUR
APOSPC2103-100G APOSPC2103-500G APOSPC2103-50G
2-Nitro-4-(trifluoromethyl)benzonitrile 99%
2-Nitro-4-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₃N₂O₂
Boiling Pt: 156…158 °C/18mmHg
Melting Pt: 44…47 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00014684
CAS Number: 778-94-9
EINECS: 212-298-1
UN: 3439
ADR: 6.1,III