4-(Trifluormethyl)nicotinonitrile 95+%
Supplier: Apollo Scientific
Warning
PC2108B-10G PC2108B-1G PC2108B-5G
APOSPC2108B-10GEA 115 EUR
APOSPC2108B-10G APOSPC2108B-1G APOSPC2108B-5G
4-(Trifluormethyl)nicotinonitrile 95+%
4-(Trifluormethyl)nicotinonitrile
Formula: C₇H₃F₃N₂
Boiling Pt: 75…77 °C/15mmHg
Storage Temperature: Ambient
MDL Number: MFCD00153107
CAS Number: 13600-43-6
UN: 3276
ADR: 6.1,III