α,α,α-Trifluoro-2,5-diaminotoluene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2,5-Diaminobenzotrifluoride, 2-(Trifluoromethyl)benzene-1,4-diamine, 2-(Trifluoromethyl)-p-phenylenediamine, 2-(Trifluoromethyl)-1,4-phenylenediamine
PC2166-25G
APOSPC2166-25GEA 178 EUR
APOSPC2166-25G
α,α,α-Trifluoro-2,5-diaminotoluene 98%
α,α,α-Trifluoro-2,5-diaminotoluene
Formula: C₇H₇F₃N₂
MW: 176.14 g/mol
Boiling Pt: 150 °C/10mmHg
Melting Pt: 54…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00036101
CAS Number: 364-13-6