α,α,α-Trifluoro-3,4-diaminotoluene 97+%
Supplier: Apollo Scientific
Warning
Synonyms: 4-(Trifluoromethyl)-1,2-phenylenediamine, 4-(Trifluoromethyl)benzene-1,2-diamine, 3,4-Diaminobenzotrifluoride, 4-(Trifluoromethyl)-o-phenylenediamine
PC2168-100G PC2168-25G PC2168-5G
APOSPC2168-100GEA 219 EUR
APOSPC2168-100G APOSPC2168-25G APOSPC2168-5G
α,α,α-Trifluoro-3,4-diaminotoluene 97+%
α,α,α-Trifluoro-3,4-diaminotoluene
Formula: C₇H₇F₃N₂
Melting Pt: 58…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00042456
CAS Number: 368-71-8
UN: 2811
ADR: 6.1,III