1,1-Dibromodifluoroethylene 97%
Supplier: Apollo Scientific
Warning
PC2244-1G PC2244-25G PC2244-5G
APOSPC2244-1GEA 230 EUR
APOSPC2244-1G APOSPC2244-25G APOSPC2244-5G
1,1-Dibromodifluoroethylene 97%
1,1-Dibromodifluoroethylene
Formula: C₂Br₂F₂
MW: 221.83
Boiling Pt: 66.2 °C/700mmHg
Density: 2.29 g/cm³ (25 °C)
Storage Temperature: Refrigerator
CAS Number: 430-85-3
UN: 2810
ADR: 6.1 III