1-Bromo-4-fluoro-2-nitrobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-5-fluoronitrobenzene
PC2256-100G PC2256-25G PC2256-5G
APOSPC2256-100GEA 58 EUR
APOSPC2256-100G APOSPC2256-25G APOSPC2256-5G
1-Bromo-4-fluoro-2-nitrobenzene 98%
1-Bromo-4-fluoro-2-nitrobenzene
Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Boiling Pt: 148…150 °C/35mmHg
Melting Pt: 39…42 °C
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00055530
CAS Number: 446-09-3
EINECS: 207-160-2