2,5-Dibromo-1-fluorobenzene 98%
Supplier: Apollo Scientific
Warning
PC2261-100G PC2261-1KG PC2261-500G
APOSPC2261-100GEA 40.6 EUR
APOSPC2261-100G APOSPC2261-1KG APOSPC2261-500G
2,5-Dibromo-1-fluorobenzene 98%
2,5-Dibromo-1-fluorobenzene
Formula: C₆H₃Br₂F
Boiling Pt: 216 °C
Melting Pt: 33…36 °C
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00010603
CAS Number: 1435-52-5