4,4'-Dibromooctafluorobiphenyl (DBOB) 99%
Supplier: Apollo Scientific
Warning
Synonyms: 4,4-Dibromooctafluorobiphenyl
PC2279-1G PC2279-25G
APOSPC2279-1GEA 62.5 EUR
APOSPC2279-1G APOSPC2279-25G
4,4'-Dibromooctafluorobiphenyl (DBOB) 99%
4,4'-Dibromooctafluorobiphenyl (DBOB)
Formula: C₁₂Br₂F₈
MW: 455.94 g/mol
Melting Pt: 113…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00000310
CAS Number: 10386-84-2
EINECS: 233-847-1
UN: 3152
ADR: 9,II