1,1-Dibromo-3,3,4,4,4-pentafluoro-2-butanone 97%
Supplier: Apollo Scientific
PC2280-1G PC2280-500MG PC2280-5G
APOSPC2280-1GEA 43.7 EUR
APOSPC2280-1G APOSPC2280-500MG APOSPC2280-5G
1,1-Dibromo-3,3,4,4,4-pentafluoro-2-butanone 97%
1,1-Dibromo-3,3,4,4,4-pentafluoro-2-butanone
Formula: C₄HBr₂F₅O
Boiling Pt: 123 °C
Storage Temperature: Ambient
CAS Number: 203302-96-9