1,2-Dibromotetrafluorobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1,2-Dibromo-3,4,5,6-tetrafluorobenzene
PC2290-25G PC2290-5G
APOSPC2290-25GEA 290 EUR
APOSPC2290-25G APOSPC2290-5G
1,2-Dibromotetrafluorobenzene 98%
1,2-Dibromotetrafluorobenzene
Formula: C₆Br₂F₄
MW: 307.87 g/mol
Boiling Pt: 198 °C
Melting Pt: 12
Density: 2.238 g/cm³ (25 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00039209
CAS Number: 827-08-7
EINECS: 212-564-7