3-Bromo-2-chlorobenzotrifluoride 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-2-chloro-3-(trifluoromethyl)benzene
PC2303-1G PC2303-5G PC2303-25G
APOSPC2303-1GEA 21.9 EUR
APOSPC2303-1G APOSPC2303-5G APOSPC2303-25G
3-Bromo-2-chlorobenzotrifluoride 98%
3-Bromo-2-chlorobenzotrifluoride
Formula: C₇H₃BrClF₃
MW: 259.45 g/mol
Boiling Pt: 75…77 °C/0.5mmHg
Storage Temperature: Ambient
MDL Number: MFCD04115994
CAS Number: 56131-47-6