3-Chloro-2-fluorobenzonitrile
Supplier: Apollo Scientific
Warning
PC2309-5G PC2309-10G
APOSPC2309-5GEA 21.9 EUR
APOSPC2309-5G APOSPC2309-10G
3-Chloro-2-fluorobenzonitrile
3-Chloro-2-fluorobenzonitrile
Formula: C₇H₃ClFN
MW: 155.56 g/mol
Melting Pt: 40…44 °C
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00042206
CAS Number: 94087-40-8
EINECS: 301-934-4
UN: 3439
ADR: 6.1 III