1,4-Dibromotetrafluorobenzene 98%
Supplier: Apollo Scientific
Warning
PC2310-5G PC2310-1G PC2310-25G
APOSPC2310-5GEA 65.4 EUR
APOSPC2310-5G APOSPC2310-1G APOSPC2310-25G
1,4-Dibromotetrafluorobenzene 98%
1,4-Dibromotetrafluorobenzene
Formula: C₆Br₂F₄
MW: 307.87 g/mol
Melting Pt: 78…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00000309
CAS Number: 344-03-6
EINECS: 206-448-5