1-Chloro-2-fluoro-3-iodobenzene 95%
Supplier: Apollo Scientific
Warning
PC2311-5G PC2311-1G PC2311-25G
APOSPC2311-5GEA 24.8 EUR
APOSPC2311-5G APOSPC2311-1G APOSPC2311-25G
1-Chloro-2-fluoro-3-iodobenzene 95%
1-Chloro-2-fluoro-3-iodobenzene
Formula: C₆H₃ClFI
Boiling Pt: 234 °C
Density: 2.008 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03094152
CAS Number: 72373-82-1