2,6-Dibromo-4-(trifluoromethoxy)aniline 97%
Supplier: Apollo Scientific
Warning
PC2330B-100G PC2330B-10G PC2330B-50G
APOSPC2330B-100GEA 34.8 EUR
APOSPC2330B-100G APOSPC2330B-10G APOSPC2330B-50G
2,6-Dibromo-4-(trifluoromethoxy)aniline 97%
2,6-Dibromo-4-(trifluoromethoxy)aniline
Formula: C₇H₄Br₂F₃NO
Boiling Pt: 65 °C/0.1mmHg
Melting Pt: 70…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00153113
CAS Number: 88149-49-9