2,6-Dibromo-4-(trifluoromethyl)aniline 95%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Amino-3,5-dibromo-α,α,α-trifluorotoluene
PC2330D-100G PC2330D-25G PC2330D-5G
APOSPC2330D-100GEA 145 EUR
APOSPC2330D-100G APOSPC2330D-25G APOSPC2330D-5G
2,6-Dibromo-4-(trifluoromethyl)aniline 95%
2,6-Dibromo-4-(trifluoromethyl)aniline
Formula: C₇H₄Br₂F₃N
MW: 318.92 g/mol
Boiling Pt: 64…65 °C/0.1mmHg
Melting Pt: 34…40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00068181
CAS Number: 72678-19-4