1,3-Difluoro-5-iodobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3,5-Difluoroiodobenzene
PC2379-25G PC2379-100G PC2379-5G
APOSPC2379-25GEA 36.4 EUR
APOSPC2379-25G APOSPC2379-100G APOSPC2379-5G
1,3-Difluoro-5-iodobenzene 98%
1,3-Difluoro-5-iodobenzene
Formula: C₆H₃F₂I
MW: 239.99 g/mol
Boiling Pt: 173…174 °C
Density: 2.004 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD03094155
CAS Number: 2265-91-0
EINECS: 607-122-9