4-Bromo-1,5-dimethyl-3-(trifluoromethyl)-1H-pyrazole 98%
Supplier: Apollo Scientific
Warning
PC2390-100MG PC2390-1G PC2390-250MG
APOSPC2390-100MGEA 91.5 EUR
APOSPC2390-100MG APOSPC2390-1G APOSPC2390-250MG
4-Bromo-1,5-dimethyl-3-(trifluoromethyl)-1H-pyrazole 98%
4-Bromo-1,5-dimethyl-3-(trifluoromethyl)-1H-pyrazole
Formula: C₆H₆BrF₃N₂
Storage Temperature: Ambient
MDL Number: MFCD04972688
CAS Number: 721402-02-4