2',4'-Dichloro-5'-fluoroacetophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(2,4-Dichloro-5-fluorophenyl)ethanone
PC2398E-100G PC2398E-25G
APOSPC2398E-100GEA 36.4 EUR
APOSPC2398E-100G APOSPC2398E-25G
2',4'-Dichloro-5'-fluoroacetophenone 98%
2',4'-Dichloro-5'-fluoroacetophenone
Formula: C₈H₅Cl₂FO
Boiling Pt: 134…135 °C/15mmHg
Melting Pt: 33…35 °C
Density: 1.425 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00077499
CAS Number: 704-10-9