1,3-Dichloro-4-fluorobenzene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 2,4-Dichlorofluorobenzene, 2,4-Dichloro-1-fluorobenzene
PC2398M-25G PC2398M-100G
APOSPC2398M-25GEA 21.9 EUR
APOSPC2398M-25G APOSPC2398M-100G
1,3-Dichloro-4-fluorobenzene 99%
1,3-Dichloro-4-fluorobenzene
Formula: C₆H₃Cl₂F
MW: 164.99 g/mol
Boiling Pt: 172…174 °C
Melting Pt: –23 °C
Density: 1.409 g/cm³ (25 °C)
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00000580
CAS Number: 1435-48-9
EINECS: 406-160-1
UN: 1993
ADR: 3,III