1,2-Dichloro-4-fluorobenzene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 3,4-Dichlorofluorobenzene
PC2400-100G PC2400-25G PC2400-500G
APOSPC2400-100GEA 88.6 EUR
APOSPC2400-100G APOSPC2400-25G APOSPC2400-500G
1,2-Dichloro-4-fluorobenzene 99%
1,2-Dichloro-4-fluorobenzene
Formula: C₆H₃Cl₂F
MW: 164.99 g/mol
Boiling Pt: 166…171 °C
Melting Pt: –1 °C
Density: 1.403 g/cm³ (25 °C)
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00018119
CAS Number: 1435-49-0
EINECS: 215-858-3