1,2,3-Trifluoro-5-nitrobenzene
Supplier: Apollo Scientific
Danger
Synonyms: 1,2,3-Trifluoro-5-nitrobenzene, 3,4,5-Trifluoronitrobenzene
PC2435-100G PC2435-25G PC2435-5G
APOSPC2435-100GEA 219 EUR
APOSPC2435-100G APOSPC2435-25G APOSPC2435-5G
1,2,3-Trifluoro-5-nitrobenzene
1,2,3-Trifluoro-5-nitrobenzene
Formula: C₆H₂F₃NO₂
Density: 1.517 g/cm³ (25 °C)
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00456803
CAS Number: 66684-58-0
EINECS: 266-447-0
UN: 2810
ADR: 6.1,III