2-Methylsulphonyl-1-(3-trifluoromethylphenyl)ethanone 95+%
Supplier: Apollo Scientific
Synonyms: 3-(Methylsulphonylacetyl)benzotrifluoride
PC2458-25G PC2458-5G PC2458-1G
APOSPC2458-25GEA 734 EUR
APOSPC2458-25G APOSPC2458-5G APOSPC2458-1G
2-Methylsulphonyl-1-(3-trifluoromethylphenyl)ethanone 95+%
2-Methylsulphonyl-1-(3-trifluoromethylphenyl)ethanone
Formula: C₁₀H₉F₃O₃S
MW: 266.24 g/mol
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00665597
CAS Number: 128306-96-7