3'-Bromo-2,2,2,4'-tetrafluoroacetophenone 97%
Supplier: Apollo Scientific
Warning
PC2465-1G PC2465-250MG PC2465-5G PC2465-25G
APOSPC2465-1GEA 429 EUR
APOSPC2465-1G APOSPC2465-250MG APOSPC2465-5G APOSPC2465-25G
3'-Bromo-2,2,2,4'-tetrafluoroacetophenone 97%
3'-Bromo-2,2,2,4'-tetrafluoroacetophenone
Formula: C₈H₃BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD08457469
CAS Number: 150698-74-1