4-Bromo-2,3,5,6-tetrafluorobenzonitrile 98%
Supplier: Apollo Scientific
Warning
PC2466-1G PC2466-10G
APOSPC2466-1GEA 21.9 EUR
APOSPC2466-1G APOSPC2466-10G
4-Bromo-2,3,5,6-tetrafluorobenzonitrile 98%
4-Bromo-2,3,5,6-tetrafluorobenzonitrile
Formula: C₇BrF₄N
MW: 253.9792 g/mol
Melting Pt: 77…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00075282
CAS Number: 17823-40-4