1,2-Dichlorohexafluorocyclopentene 97%
Supplier: Apollo Scientific
Danger
Synonyms: Perfluoro(1,2-dichlorocyclopent-1-ene)
PC2470-25G PC2470-5G
APOSPC2470-25GEA 427 EUR
APOSPC2470-25G APOSPC2470-5G
1,2-Dichlorohexafluorocyclopentene 97%
1,2-Dichlorohexafluorocyclopentene
Formula: C₅Cl₂F₆
MW: 244.95 g/mol
Boiling Pt: 90 °C
Melting Pt: –105.8 °C
Density: 1.635 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00019302
CAS Number: 706-79-6
EINECS: 211-895-4
UN: 2810
ADR: 6.1 III