5,7-Dichloro-2-(trifluoromethyl)-4-quinolinol 97%
Supplier: Apollo Scientific
Synonyms: 5,7-Dichloro-4-hydroxy-2-(trifluoromethyl)quinoline
PC2477-10G PC2477-1G PC2477-5G
APOSPC2477-10GEA 183 EUR
APOSPC2477-10G APOSPC2477-1G APOSPC2477-5G
5,7-Dichloro-2-(trifluoromethyl)-4-quinolinol 97%
5,7-Dichloro-2-(trifluoromethyl)-4-quinolinol
Formula: C₁₀H₄Cl₂F₃NO
MW: 282.0500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173851
CAS Number: 59108-13-3