3,5-Dichloro-4-iodobenzotrifluoride 97%
Supplier: Apollo Scientific
|
Warning
|
PC2477E-1G
PC2477E-5G
APOSPC2477E-1GEA
43.5
EUR
APOSPC2477E-1G
APOSPC2477E-5G
3,5-Dichloro-4-iodobenzotrifluoride 97%
3,5-Dichloro-4-iodobenzotrifluoride
Formula:
C₇H₂Cl₂F₃I Boiling Pt: 119…120 °C/20mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00084997 CAS Number: 175205-56-8 |