4-Bromo-1-fluoro-2-iodobenzene 99%
Supplier: Apollo Scientific
Warning
PC2532-100G PC2532-25G PC2532-500G
APOSPC2532-100GEA 55.1 EUR
APOSPC2532-100G APOSPC2532-25G APOSPC2532-500G
4-Bromo-1-fluoro-2-iodobenzene 99%
4-Bromo-1-fluoro-2-iodobenzene
Formula: C₆H₃BrFI
Boiling Pt: 247 °C
Density: 2.281 g/cm³ (25 °C)
Flash Pt: 103 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368751
CAS Number: 116272-41-4