1,2-Dichloro-1,1,2-trifluoro-2-iodoethane 85%
Supplier: Apollo Scientific
Warning
PC2550-100G PC2550-25G
APOSPC2550-100GEA 152 EUR
APOSPC2550-100G APOSPC2550-25G
1,2-Dichloro-1,1,2-trifluoro-2-iodoethane 85%
1,2-Dichloro-1,1,2-trifluoro-2-iodoethane
90%(min) CCl2CClFI: 10%(max) CF2ICCI2F
Formula: ClF₂CCClFI
MW: 278.83 g/mol
Boiling Pt: 100…101 °C
Density: 2.2 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00039402
CAS Number: 354-61-0