2,4-Dichlorophenyl trifluoromethyl ether 98%
Supplier: Apollo Scientific
Warning
PC2551-10G PC2551-5G
APOSPC2551-10GEA 183 EUR
APOSPC2551-10G APOSPC2551-5G
2,4-Dichlorophenyl trifluoromethyl ether 98%
2,4-Dichlorophenyltrifluoromethyl ether
Formula: C₇H₃Cl₂F₃O
Boiling Pt: 170…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00041538
CAS Number: 451-85-4