2,6-Dichloro-4-(trifluoromethyl)aniline 99%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Amino-3,5-dichloro-α,α,α-trifluorotoluene, 4-Amino-3,5-dichlorobenzotrifluoride, 2,6-Dichloro-4-trifluoromethylaniline, 2,6-Dichloro-4-(trifluoromethyl)aniline
PC2554-250G PC2554-50G PC2554-25G PC2554-500G PC2554-1KG
APOSPC2554-250GEA 102 EUR
APOSPC2554-250G APOSPC2554-50G APOSPC2554-25G APOSPC2554-500G APOSPC2554-1KG
2,6-Dichloro-4-(trifluoromethyl)aniline 99%
2,6-Dichloro-4-(trifluoromethyl)aniline
Formula: C₇H₄Cl₂F₃N
MW: 230.02 g/mol
Boiling Pt: 60…62 °C/1mmHg
Melting Pt: 34…36 °C
Density: 1.532 g/cm³ (25 °C)
Flash Pt: 88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052918
CAS Number: 24279-39-8
UN: 3077
ADR: 9+EHS,III