2,6-Dichloro-4-(trifluoromethyl)phenylhydrazine 97%
Supplier: Apollo Scientific
Warning
PC2554H-5G PC2554H-10G PC2554H-1G
APOSPC2554H-5GEA 87 EUR
APOSPC2554H-5G APOSPC2554H-10G APOSPC2554H-1G
2,6-Dichloro-4-(trifluoromethyl)phenylhydrazine 97%
2,6-Dichloro-4-(trifluoromethyl)phenylhydrazine
Formula: C₇H₅Cl₂F₃N₂
MW: 245.03 g/mol
Melting Pt: 57…61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00085001
CAS Number: 86398-94-9