2,3-Difluoro-4-nitroanisole 98%
Supplier: Apollo Scientific
Warning
PC2555-1G PC2555-25G PC2555-5G
APOSPC2555-1GEA 29 EUR
APOSPC2555-1G APOSPC2555-25G APOSPC2555-5G
2,3-Difluoro-4-nitroanisole 98%
2,3-Difluoro-4-nitroanisole
Formula: C₇H₅F₂NO₃
MW: 189.12 g/mol
Melting Pt: 96…99 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD07995055
CAS Number: 66684-59-1