Diethyl fluoromalonate 97%
Supplier: Apollo Scientific
Danger
Synonyms: 1,3-diethyl 2-fluoropropanedioate
PC2564-100G PC2564-25G PC2564-5G
APOSPC2564-100GEA 65.4 EUR
APOSPC2564-100G APOSPC2564-25G APOSPC2564-5G
Diethyl fluoromalonate 97%
Diethyl fluoromalonate
Formula: C₇H₁₁FO₄
MW: 178.16 g/mol
Boiling Pt: 121…122 °C/30mmHg
Density: 1.129 g/cm³ (25 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00009139
CAS Number: 685-88-1
EINECS: 211-684-7
UN: 1760
ADR: 8 III