2',4'-Difluoroacetanilide 99%
Supplier: Apollo Scientific
Warning
PC2569-100G PC2569-25G
APOSPC2569-100GEA 96.3 EUR
APOSPC2569-100G APOSPC2569-25G
2',4'-Difluoroacetanilide 99%
2',4'-Difluoroacetanilide
Formula: C₈H₇F₂NO
MW: 171.15 g/mol
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00032502
CAS Number: 399-36-0