Difluoroacetic acid 98%
Supplier: Apollo Scientific
Danger
PC2570-100G PC2570-25G PC2570-500G
APOSPC2570-100GEA 58 EUR
APOSPC2570-100G APOSPC2570-25G APOSPC2570-500G
Difluoroacetic acid 98%
Difluoroacetic acid
Formula: C₂H₂F₂O₂
MW: 96.03 g/mol
Boiling Pt: 134 °C
Melting Pt: –1 °C
Density: 1.526 g/cm³ (25 °C)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00004220
CAS Number: 381-73-7
EINECS: 206-839-0
UN: 3265
ADR: 8,II