3',5'-Difluoroacetophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(3,5-Difluorophenyl)ethan-1-one
PC2579-100G PC2579-25G PC2579-5G
APOSPC2579-100GEA 109 EUR
APOSPC2579-100G APOSPC2579-25G APOSPC2579-5G
3',5'-Difluoroacetophenone 98%
3',5'-Difluoroacetophenone
Formula: C₈H₆F₂O
Boiling Pt: 94…95 °C/13mmHg
Melting Pt: 34…38 °C
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00042489
CAS Number: 123577-99-1