1,3-Dibromo-5-(trifluoromethoxy)benzene
Supplier: Apollo Scientific
Warning
PC2608-100G PC2608-25G PC2608-5G
APOSPC2608-100GEA 52.2 EUR
APOSPC2608-100G APOSPC2608-25G APOSPC2608-5G
1,3-Dibromo-5-(trifluoromethoxy)benzene
1,3-Dibromo-5-(trifluoromethoxy)benzene
Formula: C₇H₃Br₂F₃O
MW: 319.90 g/mol
Boiling Pt: 83…85 °C/20mmHg
Density: 1.928 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00075280
CAS Number: 207226-31-1