3,4-Difluoroaniline 97%
Supplier: Apollo Scientific
Warning
PC2612-100G PC2612-500G
APOSPC2612-100GEA 31.9 EUR
APOSPC2612-100G APOSPC2612-500G
3,4-Difluoroaniline 97%
3,4-Difluoroaniline
Formula: C₆H₅F₂N
MW: 129.11 g/mol
Boiling Pt: 85…87 °C/30mmHg
Melting Pt: 20…22 °C
Density: 1.302 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00007761
CAS Number: 3863-11-4
EINECS: 223-381-7
UN: 2941
ADR: 6.1 III