3,4-Difluoroanisole 98%
Supplier: Apollo Scientific
Warning
PC2614H-100G PC2614H-10G PC2614H-25G PC2614H-5G
APOSPC2614H-100GEA 105 EUR
APOSPC2614H-100G APOSPC2614H-10G APOSPC2614H-25G APOSPC2614H-5G
3,4-Difluoroanisole 98%
3,4-Difluoroanisole
Formula: C₇H₆F₂O
MW: 144.12 g/mol
Boiling Pt: 139
Melting Pt: -30
Density: 1.216 g/cm³ (25 °C)
Flash Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00010730
CAS Number: 115144-40-6
UN: 1993
ADR: 3 III