2,3-Difluorobenzaldehyde 98%
Supplier: Apollo Scientific
Warning
PC2615A-100G PC2615A-10G PC2615A-50G PC2615A-25G
APOSPC2615A-100GEA 177 EUR
APOSPC2615A-100G APOSPC2615A-10G APOSPC2615A-50G APOSPC2615A-25G
2,3-Difluorobenzaldehyde 98%
2,3-Difluorobenzaldehyde
Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 70 °C/15mmHg
Density: 1.301 g/cm³ (25 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00010292
CAS Number: 2646-91-5
UN: 1989
ADR: 3,III