2,5-Difluorobenzaldehyde 98%
Supplier: Apollo Scientific
Warning
PC2615C-100G PC2615C-25G
APOSPC2615C-100GEA 144 EUR
APOSPC2615C-100G APOSPC2615C-25G
2,5-Difluorobenzaldehyde 98%
2,5-Difluorobenzaldehyde
Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 67…69 °C/17mmHg
Density: 1.308 g/cm³ (25 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00010327
CAS Number: 2646-90-4
UN: 1989
ADR: 3 III