3,5-Difluorobenzaldehyde 98%
Supplier: Apollo Scientific
Warning
PC2615F-100G PC2615F-25G
APOSPC2615F-100GEA 56.7 EUR
APOSPC2615F-100G APOSPC2615F-25G
3,5-Difluorobenzaldehyde 98%
3,5-Difluorobenzaldehyde
Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 61…63 °C/43mmHg
Melting Pt: 17 °C
Density: 1.296 g/cm³ (25 °C)
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00010329
CAS Number: 32085-88-4
UN: 1989
ADR: 3 III