3,4-Difluorobenzamide 97%
Supplier: Apollo Scientific
Warning
PC2619-100G PC2619-10G PC2619-50G
APOSPC2619-100GEA 114 EUR
APOSPC2619-100G APOSPC2619-10G APOSPC2619-50G
3,4-Difluorobenzamide 97%
3,4-Difluorobenzamide
Formula: C₇H₅F₂NO
Melting Pt: 129…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00015549
CAS Number: 85118-04-3
EINECS: 285-656-8