4-Fluoro-3-(methylsulphonyl)benzoic acid 99%
Supplier: Apollo Scientific
Warning
PC2628-1G PC2628-5G
APOSPC2628-1GEA 80 EUR
APOSPC2628-1G APOSPC2628-5G
4-Fluoro-3-(methylsulphonyl)benzoic acid 99%
4-Fluoro-3-(methylsulphonyl)benzoic acid
Formula: C₈H₇FO₄S
Melting Pt: 215…216 °C
Storage Temperature: Ambient
MDL Number: MFCD08458082
CAS Number: 158608-00-5